Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DDTGSTVSEK EDIKAKNVSS YEKTQSYPTD WSDDESNNPF SSTDANGDSN PFDDDATSGT E
PACSIN 2, a 486 AA phosphoprotein is a novel member of the PACSIN family of cytoplasmic adapter proteins. It is an isoform of PACSIN with a CDC15 N-terminal domain, a C-terminal SH3 domain, 3 conserved regions specific to the PACSIN family, and 3 asn-pro-phe (NPF) motifs, which potentially bind to EH domains. This protein is known to display a ubiquitous expression pattern with the highest levels of expression in brain. Reports suggest that PACSIN 2 participates in the organization of the actin cytoskeleton, the regulation of vesicular traffic. It also participates in endocytic internalization and endocytic receptor recycling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytoplasmic phosphoprotein PACSIN2; Protein kinase C and casein kinase substrate in neurons protein 2; RP3-437M21.1; SdpII; syndapin 2; syndapin II; Syndapin-2; Syndapin-II; syndapin2
Gene Aliases: PACSIN2; SDPII
UniProt ID: (Human) Q9UNF0
Entrez Gene ID: (Human) 11252
Molecular Function: actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support