Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: RETSAVLERK ISTRQSREEL IRRGVLKELP DQDGDVTVNF ENSNGHMIPI GEESTREENV VKSEEGNGSV SEKTPPLEEQ AEDKKENT
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Phosphatase and actin regulator 2
Gene Aliases: C6orf56; KIAA0680; PHACTR2
UniProt ID: (Human) O75167, (Rat) P62025
Entrez Gene ID: (Human) 9749, (Rat) 308291
Molecular Function: phosphatase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support