Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
82% identical to rat and 81% identical to mouse. >30% identity with DMPK.
1 µg/mL of MA5-45655 was sufficient for detection of PINK1 in 20 µg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 50kDa.
This antibody was formerly sold as clone S4-15.
PINK1 (PTEN-induced putative kinase 1) protects from mitochondrial dysfunction in cells by conferring different autophosphorylation activity to mitochondrial genes. The kinase domain of PINK1 appears to be critical in its protective effects. Mitochondrial proteins are critical for the regulation of cellular energy and adaptation to stress. Particularly in brain cells, mitochondrial demand is extraordinarily high, in order to accommodate aerobic and anaerobic support of high energy processes. When mitochondrial structure and/or function is disrupted in the brain, neurodegenerative diseases like Parkinson's disease are triggered.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BRPK; FLJ27236; PINK 1; PINK-1; protein kinase BRPK; PTEN-induced putative kinase protein 1; Serine/threonine-protein kinase PINK1, mitochondrial
Gene Aliases: 1190006F07Rik; AU042772; AW557854; BRPK; mFLJ00387; PARK6; PINK1
UniProt ID: (Human) Q9BXM7, (Mouse) Q99MQ3
Entrez Gene ID: (Human) 65018, (Rat) 298575, (Mouse) 68943
Molecular Function: non-receptor serine/threonine protein kinase protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support