Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEPKRIREGY LVKKGSVFNT WKPMWVVLLE DGIEFYKKKS DNSPKGMIPL KGSTLTSPCQ DFGKRMFVFK ITTTKQQDHF FQAAFLEERD AWVRDINKAI KCIEGGQKFA RKSTRRSIRL PETIDLGALY LSMKDTEKGI KELNLEKDKK IFNHCFTGNC VIDWLVSNQS VRNRQEGLMI ASSLLNEGYL QPAGDMSKSA VDGTAENPFL DNPDAFYYFP DSGFFCEENS SDDDVILKEE FRGVIIKQGC LLKQGHRRKN WKVRKFILRE DPAYLHYYDP AGAEDPLGAI HLRGCVVTSV ESNSNGRKSE EENLFEIITA DEVHYFLQAA TPKERTEWIK AIQMASRTGK
Major protein kinase C substrate of platelets.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ27168; p47; Platelet 47 kDa protein; Pleckstrin
Gene Aliases: P47; PLEK
UniProt ID: (Human) P08567
Entrez Gene ID: (Human) 5341
Molecular Function: cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support