Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTSSGPGPRF LLLLPLLLPP AASASDRPRG RDPVNPEKLL VITVATAETE GYLRFLRSAE FFNYTVRTLG LGEEWRGGDV ARTVGGGQKV RWLKKEMEKY ADREDMIIMF VDSYDVILAG SPTELLKKFV QSGSRLLFSA ESFCWPEWGL AEQYPEVGTG KRFLNSGGFI GFATTIHQIV RQWKYKDDDD DQLFYTRLYL DPGLREKLSL NLDHKSRIFQ NLNGALDEVV LKFDRNRVRI RNVAYDTLPI VVHGNGPTKL QLNYLGNYVP NGWTPEGGCG FCNQDRRTLP GGQPPPRVFL AVFVEQPTPF LPRFLQRLLL LDYPPDRVTL FLHNNEVFHE PHIADSWPQL QDHFSAVKLV GPEEALSPGE ARDMAMDLCR QDPECEFYFS LDADAVLTNL QTLRILIEEN RKVIAPMLSR HGKLWSNFWG ALSPDEYYAR SEDYVELVQR KRVGVWNVPY ISQAYVIRGD TLRMELPQRD VFSGSDTDPD MAFCKSFRDK GIFLHLSNQH EFGRLLATSR YDTEHLHPDL WQIFDNPVDW KEQYIHENYS RALEGEGIVE QPCPDVYWFP LLSEQMCDEL VAEMEHYGQW SGGRHEDSRL AGGYENVPTV DIHMKQVGYE DQWLQLLRTY VGPMTESLFP GYHTKARAVM NFVVRYRPDE QPSLRPHHDS STFTLNVALN HKGLDYEGGG CRFLRYDCVI SSPRKGWALL HPGRLTHYHE GLPTTWGTRY IMVSFVDP
Forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: lysine hydroxylase 3; lysyl hydroxlase 3; lysyl hydroxylase 3; Multifunctional procollagen lysine hydroxylase and glycosyltransferase LH3; procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3
Gene Aliases: LH3; PLOD3
UniProt ID: (Human) O60568
Entrez Gene ID: (Human) 8985
Molecular Function: protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support