Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAACRYCCSC LRLRPLSDGP FLLPRRDRAL TQLQVRALWS SAGSRAVAVD LGNRKLEISS GKLARFADGS AVVQSGDTAV MVTAVSKTKP SPSQFMPLVV DYRQKAAAAG RIPTNYLRRE VGTSDKEILT SRIIDRSIRP LFPAGYFYDT QVLCNLLAVD GVNEPDVLAI NGASVALSLS DIPWNGPVGA VRIGIIDGEY VVNPTRKEMS SSTLNLVVAG APKSQIVMLE ASAENILQQD FCHAIKVGVK YTQQIIQGIQ QLVKETGVTK RTPQKLFTPS PEIVKYTHKL AMERLYAVFT DYEHDKVSRD EAVNKIRLDT EEQLKEKFPE ADPYEIIESF NVVAKEVFRS IVLNEYKRCD GRDLTSLRNV SCEVDMFKTL HGSALFQRGQ TQVLCTVTFD SLESGIKSDQ VITAINGIKD KNFMLHYEFP PYATNEIGKV TGLNRRELGH GALAEKALYP VIPRDFPFTI RVTSEVLESN GSSSMASACG GSLALMDSGV PISSAVAGVA IGLVTKTDPE KGEIEDYRLL TDILGIEDYN GDMDFKIAGT NKGITALQAD IKLPGIPIKI VMEAIQQASV AKKEILQIMN KTISKPRASR KENGPVVETV QVPLSKRAKF VGPGGYNLKK LQAETGVTIS QVDEETFSVF APTPSAMHEA RDFITEICKD DQEQQLEFGA VYTATITEIR DTGVMVKLYP NMTAVLLHNT QLDQRKIKHP TALGLEVGQE IQVKYFGRDP ADGRMRLSRK VLQSPATTVV RTLNDRSSIV MGEPISQSSS NSQ
PNPT1 (polyribonucleotide nucleotidyltransferase 1, mitochondirla) is a RNA-binding protein implicated in numerous RNA metabolic processes. It catalyzes the phosphorolysis of single-stranded polyribonucleotides processively in the 3'-to-5' direction. PNPT1 is a component of the mitochondrial degradosome (mtEXO) complex that degrades 3' overhang double-stranded RNA with a 3'-to-5' directionality in an ATP-dependent manner. It is required for correct processing and polyadenylation of mitochondrial mRNAs. It plays a role as a cytoplasmic RNA import factor, in mitochondrial morphogenesis and respiration, regulation of expression of the electron transport chain, regulation of the stability of specific mature miRNAs in melanoma cells, and RNA cell surveilance. Mutations of the gene can result in combined oxidative phosphorylation deficiency 13 (COXPD13) and deafness, autosomal recessive, 70 (DFNB70).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'-5' RNA exonuclease OLD35; DKFZp762K1914; old 35; PNPase 1; PNPase old-35; Polynucleotide phosphorylase 1; Polynucleotide phosphorylase-like protein; Polyribonucleotide nucleotidyltransferase 1, mitochondrial
Gene Aliases: 1200003F12Rik; COXPD13; DFNB70; old-35; OLD35; PNPASE; PNPT1; Pnptl1
UniProt ID: (Human) Q8TCS8, (Mouse) Q8K1R3
Entrez Gene ID: (Human) 87178, (Mouse) 71701
Molecular Function: nucleotidyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support