Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MSALCWGRGA AGLKRALRPC GRPGLPGKEG TAGGVCGPRR SSSASPQEQD QDRRKDWGHV ELLEVLQARV RQLQAESVSE VVVNRVDVAR LPECGSGDGS
This gene encodes a mitochondrial DNA-directed RNA polymerase. The gene product is responsible for mitochondrial gene expression as well as for providing RNA primers for initiation of replication of the mitochondrial genome. Although this polypeptide has the same function as the three nuclear DNA-directed RNA polymerases, it is more closely related to RNA polymerases of phage and mitochondrial polymerases of lower eukaryotes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA-directed RNA polymerase, mitochondrial; MtRPOL; polymerase (RNA) mitochondrial (DNA directed)
Gene Aliases: APOLMT; h-mtRPOL; MTRNAP; MTRPOL; POLRMT
UniProt ID: (Human) O00411
Entrez Gene ID: (Human) 5442
Molecular Function: DNA-directed RNA polymerase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support