Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes an alpha isoform of the catalytic subunit.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PP2A-alpha; PP2A-beta; protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform; protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform; protein phosphatase 2, catalytic subunit, alpha isoform; protein phosphatase 2, catalytic subunit, alpha isozyme; protein phosphatase 2, catalytic subunit, beta isoform; protein phosphatase 2, catalytic subunit, beta isozyme; protein phosphatase 2A catalytic alpha subunit; protein phosphatase 2A catalytic subunit, beta isoform; protein phosphatase 2a, catalytic subunit, alpha isoform; protein phosphatase 2a, catalytic subunit, beta isoform; protein phosphatase type 2A catalytic subunit; protein phosphatase-2A-alpha; protein phosphatase-2A-beta; Replication protein C; RP C; RP-C; serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform; serine/threonine protein phosphatase 2A, catalytic subunit, beta isoform; Serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform; Serine/threonine-protein phosphatase 2A catalytic subunit beta isoform; testicular tissue protein Li 146
Gene Aliases: AI115466; D8Ertd766e; PP2A; Pp2a1; Pp2a2; PP2Abeta; PP2Ac; PP2CA; PP2Calpha; PP2CB; PPP2CA; PPP2CB; R75353; RP-C
UniProt ID: (Human) P67775, (Human) P62714, (Mouse) P63330, (Rat) P63331, (Rat) P62716, (Mouse) P62715
Entrez Gene ID: (Human) 5515, (Human) 5516, (Mouse) 19052, (Rat) 24672, (Rat) 24673, (Mouse) 19053
Molecular Function: protein phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support