Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine and Orangutan
Protein phosphatase methylesterase-1 catalyzes the demethylation of the protein phosphatase-2A catalytic subunit.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PME-1; Protein phosphatase methylesterase 1; protein phosphatase methylesterase-1; testicular secretory protein Li 39
Gene Aliases: 1110069N17Rik; 2700017M01Rik; PME-1; PME1; PP2593; PPME1; PRO0750
UniProt ID: (Human) Q9Y570, (Mouse) Q8BVQ5
Entrez Gene ID: (Human) 51400, (Mouse) 72590
Molecular Function: protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support