Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene was identified based on its similarity with the mouse counterpart. Studies of the mouse counterpart suggest that the expression of this gene may be regulated during oocyte maturation and preimplantation following zygotic gene activation. Alternatively spliced transcript variants encoding distinct isoforms have been observed. Naturally occurring read-through transcription occurs between this locus and the neighboring locus HSPE1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2C4D; Class II mMOB1; MOB-like protein phocein; Mob1 homolog 3; MOB1, Mps One Binder kinase activator-like 3; Mob3; Mps One Binder kinase activator-like 3; Phocein; phocein, Mob-like protein; Preimplantation protein 3
Gene Aliases: 2610109B12Rik; 2C4D; CGI-95; MOB1; MOB3; MOB4; Mobkl1; MOBKL3; phocein; PHOCN; PREI3
UniProt ID: (Human) Q9Y3A3, (Rat) Q9QYW3, (Mouse) Q6PEB6
Entrez Gene ID: (Human) 25843, (Rat) 171050, (Mouse) 19070
Molecular Function: kinase activator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support