Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
AA Sequence of recombinant protein: AVITGACERD VQCGAGTCCA ISLWLRGLRM CTPLGREGEE CHPGSHKVPF FRKRKHHTCP CLPNLLCSRF PDGRYRCSMD LKNINF.
Preparation: Produced from sera of goats immunized with highly pure Recombinant Human EG-VEGF. Anti-Human EG-VEGF-specific antibody was purified by affinity chromatography and then biotinylated.
Sandwich ELISA: To detect hEG-VEGF by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human EG-VEGF (500-P188G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hEG-VEGF.
Western Blot: To detect Human EG-VEGF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human EG-VEGF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
500-P188GBT-1MG will be provided as 2 x 500 µg
Endocrine gland-derived vascular endothelial growth factor induces proliferation, migration, and fenestration in capillary endothelial cells derived from endocrine glands. Its expression is induced by hypoxia and is restricted to the steroidogenic glands. Its expression is often complementary to the expression of VEGF, suggesting that these molecules function in a coordinated manner.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: black mamba toxin-related protein; EG-VEGF; Endocrine-gland-derived vascular endothelial growth factor; Mambakine; Prokineticin-1
Gene Aliases: EGVEGF; PK1; PRK1; PROK1; UNQ600/PRO1186
UniProt ID: (Human) P58294
Entrez Gene ID: (Human) 84432
Molecular Function: peptide hormone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support