Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YARQAINDGV TSTEELSITR DCELYRALNM HYNKANDFEV PERFLEVAQI TLREFFNAII AGKDVDPSWK KAIYKVICKL DSEVPEIFKS PNCLQELLHE
The expression pattern of the Prox1 homeo box gene suggests that it has a role in a variety of embryonic tissues, including lens. Analysis of mRNA reveals that Prox mRNA is present in many different human tissues and that lens demonstrated the highest level. Homozygous Prox1-null mice die at midgestation from multiple developmental defects, and a targeted effect on lens development has been reported. Prox1 inactivation caused abnormal cellular proliferation, downregulated expression of the cell cycle inhibitors Cdkn1b and Cdkn1c, misexpression of E-cadherin, and excessive apoptosis. Consequently, mutant lens cells failed to polarize and elongate properly, resulting in a hollow lens. The Prox1 gene is expressed in a subpopulation of endothelial cells that by budding and sprouting give rise to the lymphatic system. Prox1 appears to be a specific and required regulator of the development of the lymphatic system. Prox1 also has been document to be required for hepatocyte migration in the mouse. Loss of Prox1 results in a smaller liver with a reduced population of clustered hepatocytes. The homeodomain protein Prox1 regulates the egress of progenitor cells from the cell cycle in the embryonic mouse retina. Cells lacking Prox1 are less likely to stop dividing, and ectopic expression of Prox1 forces progenitor cells to exit the cell cycle. Prox1 acts as a key participant in progenitor-cell proliferation and cell-fate determination in the vertebrate retina.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox prospero-like protein PROX1; Prospero homeobox protein 1; prospero-related homeobox 1; PROX-1
Gene Aliases: PROX1
UniProt ID: (Human) Q92786
Entrez Gene ID: (Human) 5629
Molecular Function: homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support