Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-34860 detects HSPG2 from porcine, mouse, and human samples.
MA1-34860 has been successfully used in immunofluorescence and immunohistochemistry (frozen tissue) applications. Tonsil is suggested as a positive control.
The MA1-34860 immunogen is mouse EHS Laminin.
Perlecan is a major heparan-sulfate proteoglycan (HSPG) found within all basement membranes and cell surfaces. Because of its strategic location and ability to store and protect growth factors, perlecan has been strongly implicated in the control of tumor cell growth and metastatic behavior. Perlecan possesses angiogenic and growth-promoting attributes primarily by acting as a coreceptor for basic fibroblast growth factor (FGF-2). Suppression of perlecan causes substantial inhibition of neoplastic growth and neovascularization. Thus, perlecan is a potent inducer of neoplasm growth and angiogenesis in vivo and therapeutic interventions targeting this key modulator of tumor progression may improve neoplastic treatment.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Basement membrane-specific heparan sulfate proteoglycan core protein; Endorepellin; endorepellin (domain V region); HSPG; LG3 peptide; Perlecan; perlecan proteoglycan; PLC; RP11-132G19.2
Gene Aliases: HSPG2
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support