Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aplysia Ras-related homologs (ARHs), also called Rho genes, belong to the RAS gene superfamily encoding small guanine nucleotide exchange (GTP/GDP) factors. The ARH proteins may be kept in the inactive, GDP-bound state by interaction with GDP dissociation inhibitors, such as ARHGDIA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: arhgdia {ECO:0000312|RGD:1359547}; epididymis secretory sperm binding protein Li 47e; GDI-1; GDP-dissociation inhibitor, aplysia RAS-related 1; MGC117248; Rho GDI 1; rho GDI 1 {ECO:0000250|UniProtKB:P19803}; Rho GDI alpha; Rho GDP dissociation inhibitor (GDI) alpha; Rho GDP-dissociation inhibitor 1; rho GDP-dissociation inhibitor 1 {ECO:0000250|UniProtKB:P19803}; Rho-GDI alpha; rho-GDI alpha {ECO:0000250|UniProtKB:P19803}
Gene Aliases: ARHGDIA; C87222; Gdi1; GDIA1
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support