Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of CD32 only when phosphorylated at Tyr288.
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD16; CD32; CD32 receptor 2; CD32 receptor 2a; CDw32; Fc fragment of IgG low affinity IIa receptor for (CD32); Fc fragment of IgG, low affinity II, receptor for (CD32); Fc fragment of IgG, low affinity IIa, receptor (CD32); Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity IIb, receptor for (CD32); Fc fragment of IgG, low affinity IIc, receptor for (CD32); Fc gamma receptor IIa; Fc gamma receptor IIb; Fc gamma receptor IIC; Fc gamma RIIb; Fc receptor, IgG, low affinity IIb; Fc receptor, IgG, low affinity III; FC-gamma RII; Fc-gamma RII-a; Fc-gamma RII-b; Fc-gamma RII-c; Fc-gamma RIII; Fc-gamma-RIIa; Fc-gamma-RIIb; Fc-gamma-RIIc; FcRII; fcRII-a; fcRII-b; FcRIII; FGFR2B; IgG Fc receptor II beta; IgG Fc receptor II-a; IgG Fc receptor II-b; IgG Fc receptor II-c; IgG Fc receptor III; Immunoglobulin G Fc receptor II; immunoglobulin G Fc receptor II-c; Low affinity immunoglobulin gamma Fc region receptor II; Low affinity immunoglobulin gamma Fc region receptor II-a; Low affinity immunoglobulin gamma Fc region receptor II-b; Low affinity immunoglobulin gamma Fc region receptor II-c; Low affinity immunoglobulin gamma Fc region receptor III; MGC23887; MGC30032; RP11-5K23.6
Gene Aliases: CD32; CD32A; CD32B; CD32C; CDW32; FCG2; FcgammaRIIB2; FcGR; FCGR2; FCGR2A; FCGR2A1; FCGR2B; FCGR2C; Fcgr3; Fcgr3a; FCRII; FCRIIC; IGFR2
UniProt ID: (Human) P12318, (Human) P31994, (Human) P31995, (Rat) P27645, (Rat) Q63203
Entrez Gene ID: (Human) 2212, (Human) 2213, (Human) 9103, (Rat) 116591, (Rat) 289211
Molecular Function: immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support