Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
This antibody detects endogenous protein at a molecular weight of 38 kDa.
Purity is >95% by SDS-PAGE.
CREB (Cyclic AMP response element binding protein) is a 43 kDa basic/leucine zipper transcription factor that binds the cyclic AMP response element (CRE) and activates transcription in response to a variety of extracellular signals including neurotransmitters, hormones, membrane depolarization, and growth and neurotrophic factors. Activation of CREB is dependent upon the phosphorylation of serine 133. Phosphorylation of CREB occurs via p44/42 MAP kinase and p90RSK, and via p38 MAP kinase and MSK1. Although CREB will bind DNA independent of its phosphorylation state, only the phosphorylated form is competent as a transcription factor. CREB binding protein (CBP), a transcriptional coactivator that directly interacts with CREB, binds to CREB in the region of serine 133. CREB is involved in different cellular processes including the synchronization of circadian rhythmicity, differentiation of adipose cells, and learning and memory. In humans, the gene is located on the q arm of chromosome 2. Diseases associated with CREB dysfunction include Alzheimer’s Disease, histiocytoma and soft tissue melanoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: active transcription factor CREB; cAMP response element binding protein 1; cAMP-response element-binding protein-1; cAMP-responsive element-binding protein 1; CREB-1; cyclic adenosine 3',5'-monophosphate response element-binding protein CREB; Cyclic AMP-responsive element-binding protein 1; htCREB; MGC9284; OTTHUMP00000206661; OTTHUMP00000206662; OTTHUMP00000206667; transactivator protein; Y protein
Gene Aliases: 2310001E10Rik; 3526402H21Rik; AV083133; CREB; CREB-1; CREB1
UniProt ID: (Human) P16220, (Mouse) Q01147, (Rat) P15337
Entrez Gene ID: (Human) 1385, (Mouse) 12912, (Rat) 81646
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support