Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Antibody detects endogenous levels of CTDSPL2 only when phosphorylated at Ser9.
Probable phosphatase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase like 2; CTD small phosphatase-like protein 2; CTDSP-like 2
Gene Aliases: CTDSPL2; D2Ertd485e; HSPC058; HSPC129
UniProt ID: (Human) Q05D32, (Mouse) Q8BG15
Entrez Gene ID: (Human) 51496, (Mouse) 329506
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support