Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
It is not recommended to aliquot this antibody.
RPS6KA2 (RSK3) serine/threonine kinase is a member of the ribosomal S6 kinase (RSK) family. It is believed that RPS6KA2 is a signal-transducing intermediate in the cellular response to several growth factors. It is found in many tissues, but in higher levels in lung and skeletal muscle. The p90 ribosomal S6 kinases (RSK)1-4 are downstream members of the extracellular signal-regulated kinase (ERK)/MAPK cascade. The loss of RSK2 activity in humans leads to Coffin-Lowry syndrome, which is characterized by mental retardation and growth deficits and may have a tonic regulation on G-protein coupled signaling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 90 kDa ribosomal protein S6 kinase 2; MAP kinase-activated protein kinase 1c; MAPK-activated protein kinase 1c; MAPKAP kinase 1C; MAPKAPK-1c; mitogen-activated protein kinase-activated protein kinase 1C; p90-RSK 2; p90RSK2; pp90RSK3; Protein-tyrosine kinase Mpk-9; ribosomal protein S6 kinase alpha 2; Ribosomal protein S6 kinase alpha-2; ribosomal protein S6 kinase, 90kD, polypeptide 2; ribosomal protein S6 kinase, 90kDa, polypeptide 2; Ribosomal S6 kinase 3; RSK-3; S6K-alpha 2; S6K-alpha-2
Gene Aliases: 90kDa; D17Wsu134e; HU-2; MAPKAPK1C; p90-RSK3; p90rsk; p90RSK2; pp90rsk; pp90RSK3; Rps6ka-rs1; RPS6KA2; RSK; RSK3; S6K-alpha; S6K-alpha2
UniProt ID: (Human) Q15349, (Mouse) Q9WUT3
Entrez Gene ID: (Human) 6196, (Mouse) 20112, (Rat) 117269
Molecular Function: protein modifying enzyme non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support