Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody stains human Prominin 2-transfected cells but not irrelevant transfectants.
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a member of the prominin family of pentaspan membrane glycoproteins. The encoded protein localizes to basal epithelial cells and may be involved in the organization of plasma membrane microdomains. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: h prominin-2; hPROML2; PROM-2; Prominin-2; Prominin-like protein 2; prominin-related protein
Gene Aliases: PROM2; PROML2; UNQ2521/PRO6014
UniProt ID: (Human) Q8N271
Entrez Gene ID: (Human) 150696
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support