Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is predicted to react with bovine based on sequence homology.
RAB13 can participate in polarized transport, in the assembly and/or the activity of tight junctions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell growth-inhibiting gene 4 protein; growth-inhibiting gene 4 protein; RAS-associated protein RAB13; Ras-related protein Rab-13
Gene Aliases: 0610007N03Rik; B230212B15Rik; GIG4; RAB13
UniProt ID: (Human) Q96GU4, (Mouse) Q9DD03
Entrez Gene ID: (Human) 5872, (Mouse) 68328
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support