Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A 1:1,000 dilution of MA5-45968 was sufficient for detection of RAB1A in 10 ug (A549, MEF); 20 ug (Mouse Brain) lysates by ECL immunoblot analysis.|Detects approximately 23 kDa. Cross-reacts with RAB1B and RAB1C.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a member of the L18AE family of ribosomal proteins that is a component of the 60S subunit. The encoded protein may play a role in viral replication by interacting with the hepatitis C virus internal ribosome entry site (IRES). This gene is co-transcribed with the U68 snoRNA, located within the third intron. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed throughout the genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP564B163; GTP binding protein Rab1a; Rab GTPase YPT1 homolog; RAB1, member RAS oncogene family; Ras-related protein Rab-1A; ras-related YPT1 protein; YPT1-related protein
Gene Aliases: Gtbp; mKIAA3012; Rab-1; RAB1; RAB1A; YPT1
UniProt ID: (Human) P62820, (Mouse) P62821
Entrez Gene ID: (Human) 5861, (Mouse) 19324
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support