Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | - |
Immunohistochemistry (IHC) |
- | View 1 publication 1 publication |
Product Specifications | |
---|---|
Species Reactivity |
Human, Rat |
Published species |
Human |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
336 |
Immunogen |
Synthetic peptide corresponding to the N-terminal region of human RARB conjugated to KLH |
Conjugate |
Unconjugated |
Form |
Liquid |
Purification |
Protein G |
Storage buffer |
0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol |
Contains |
no preservative |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
RRID |
AB_560887 |
This antibody is predicted to react with canine, guinea pig, mouse and non-human primate based on 100% sequence homology.
This antibody is specific for the ~48 kDa RXR-b protein.
Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HBV-activated protein; hepatitis B virus activated protein; Nuclear receptor subfamily 1 group B member 2; RAR beta; RAR-b; RAR-beta; RAR-Beta2; RAR-Beta4; RAR-epsilon; retinoic acid; retinoic acid receptor; Retinoic acid receptor beta; retinoic acid receptor, beta polypeptide
Gene Aliases: HAP; MCOPS12; NR1B2; RARB; RARbeta1; RRB2
UniProt ID: (Human) P10826
Entrez Gene ID: (Human) 5915, (Rat) 24706
Molecular Function:
C4 zinc finger nuclear receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support