Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: QKVSMHYSDP KPKINEDWLC NKCGVQNFKR REKCFKCGVP KSEAEQKLPL GTRLDQQTLP LGGRELSQGL LPLPQPYQAQ GVLASQALSQ GSEPSSENAN DTIILRNL
The protein encoded by this gene contains RNA recognition motif found in a variety of RNA binding proteins, including various hnRNP proteins, proteins implicated in regulation of alternative splicing, and protein components of snRNPs. In vitro studies showed that the rat homolog bound to RNA homopolymers, with a preference for G and U polyribonucleotides. This gene is part of a gene cluster on chromosome Xp11.23, and its 3' end lies within 20 kb upstream of UBE1. Two transcript variants encoding different isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G patch domain-containing protein 9; rna-binding; RNA-binding motif protein 10; RNA-binding protein 10; RNA-binding protein S1-1; S1-1
Gene Aliases: DXS8237E; GPATC9; GPATCH9; KIAA0122; RBM10; S1-1; TARPS; ZRANB5
UniProt ID: (Human) P98175
Entrez Gene ID: (Human) 8241
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support