Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
RRAGD is a monomeric guanine nucleotide-binding protein, or G protein. By binding GTP or GDP, small G proteins act as molecular switches in numerous cell processes and signaling pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Rag D; Rag D protein; ragD; Ras-related GTP binding D; Ras-related GTP-binding protein D; RRAGD
Gene Aliases: 5730543C08Rik; AI467523; bA11D8.2.1; C030003H22Rik; D4Ertd174e; RAGD; RRAGD
UniProt ID: (Human) Q9NQL2, (Mouse) Q7TT45
Entrez Gene ID: (Human) 58528, (Mouse) 52187
Molecular Function: small GTPase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support