Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sheep anti Rat albumin antibody recognizes rat albumin, a soluble protein which constitutes approximately 50% of blood serum protein, acting as a regulator of blood colloidal osmotic pressure and hence blood volume.
Albumin is a soluble protein which constitutes approximately 50% of blood serum protein, and acts as a regulator of blood colloidal osmotic pressure and blood volume. Albumin is also an important transporter of substances within the bloodstream, such as fatty acids, thyroid hormones, metal ions and steroids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Albumin; BSA; serum albumin
Gene Aliases: ALB; Alb1; Albza
UniProt ID: (Bovine) P02769, (Rat) P02770
Entrez Gene ID: (Bovine) 280717, (Rat) 24186
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support