Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
It is not recommended to aliquot this antibody.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a cytoplasmic ribosomal protein that is a component of the 40S subunit. The protein belongs to the S6E family of ribosomal proteins. It is the major substrate of protein kinases in the ribosome, with subsets of five C-terminal serine residues phosphorylated by different protein kinases. Phosphorylation is induced by a wide range of stimuli, including growth factors, tumor-promoting agents, and mitogens. Dephosphorylation occurs at growth arrest. The protein may contribute to the control of cell growth and proliferation through the selective translation of particular classes of mRNA. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40S ribosomal protein S6; CG10944-PA; CG10944-PB; hemocytes enlarged; LD31286p; lethal(1)aberrant immune response[8]; lethal-(1)-haemocytes enlarged; lethal-aberrant-immune-response-8; minute; minute (1) 7BC; OTTHUMP00000021121; OTTHUMP00000021122; Phosphoprotein NP33; RP11-513M16.6; RpS6-PA; RpS6-PB; RS6; S6 ribosomal protein; Small ribosomal subunit protein eS6
Gene Aliases: (Rp)S6; air8; air[8]; anon-WO02059370.61; CG10944; Dmel\CG10944; Dmel_CG10944; DS6; hen; l(1)air8; l(1)air[8]; l(1)hen; M(1)7BC; M(1)7C; OK/SW-cl.2; pp30; Rp S6; RPS6; S6; S6R
UniProt ID: (Human) P62753, (Fruit fly) P29327, (Mouse) P62754, (Rat) P62755
Entrez Gene ID: (Human) 6194, (Fruit fly) 31700, (Mouse) 20104, (Rat) 29304
Molecular Function: ribosomal protein translational protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support