Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
By Western blot this antibody detects a band at ~12 kDa.
The serum amyloid A family comprises a number of differentially expressed apolipoproteins, acute-phase SAA1 and SAA2, the former being the major component in plasma and constitutive SAAs. Although the liver is the primary site of synthesis of both SAA types extrahepatic production has been reported. The in vivo concentrations increase by as much as 1000 fold during inflammation. Several studies have expressed its importance in the diagnosis and monitoring of various diseases. Pathological SAA values are often detected in association with normal CRP concentrations; SAA rises earlier and more sharply than CRP. Recently, a broader view of SAA expression and function has been emerging.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: amyloid a; Amyloid fibril protein AA; Amyloid protein A; MGC111216; SAA; serum amyloid A protein; Serum amyloid A-1 protein; Serum amyloid A1 protein; Serum amyloid protein A(2-102); Serum amyloid protein A(2-103); Serum amyloid protein A(2-104); Serum amyloid protein A(3-104); Serum amyloid protein A(4-101); Serum Amyloid-A; tumor protein p53 inducible protein 4
Gene Aliases: PIG4; SAA; SAA1; SAA2; TP53I4
UniProt ID: (Human) P0DJI8
Entrez Gene ID: (Human) 6288
Molecular Function: apolipoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support