Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MDKEYVGFAA LPNQLHRKSV KKGFDFTLMV AGESGLGKST LINSLFLTNL YEDRQVPEAS ARLTQTLAIE RRGVEIEEGG VKVKLTLVDT PGFGDSVDCS DCWLPVVKFI EEQFEQYLRD ESGLNRKNIQ DSRVHCCLYF ISPFGRGLRP LDVAFLRAVH EKVNIIPVIG KADALMPQET QALKQKIRDQ LKEEEIHIYQ FPECDSDEDE DFKRQDAEMK ESIPFAVVGS CEVVRDGGNR PVRGRRYSWG TVEVENPHHC DFLNLRRMLV QTHLQDLKEV THDLLYEGYR ARRLQSLARP GARDRASRSK LSRQSATEIP LPMLPLADTE KLIREKDEEL RRMQEMLEKM QAQMQRSQAQ GEQSDAL
Septin 1 (SEPT1) is a member of the septin family of GTPases. Members of this family are required for cytokinesis and the maintenance of cellular morphology. This gene encodes a protein that can form homo- and heterooligomeric filaments, and may contribute to the formation of neurofibrillary tangles in Alzheimer's disease. Alternatively spliced transcript variants have been found but the full-length nature of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-Sep; differentiation 6; differentiation 6 (deoxyguanosine triphosphate triphosphohydrolase); LARP; MGC20394; Peanut-like protein 3; Septin-1; Serologically defined breast cancer antigen NY-BR-24
Gene Aliases: DIFF6; LARP; PNUTL3; SEP1; SEPT1; SEPTIN1
UniProt ID: (Human) Q8WYJ6
Entrez Gene ID: (Human) 1731
Molecular Function: cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support