Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDPLRRSPSP CLSSQPSSPS TPPCEMLGPV GIEAVLDQLK IKAMKMGFEF NIMVVGQSGL GKSTMVNTLF KSKVWKSNPP GLGVPTPQTL QLHSLTHVIE EKGVKLKLTV TDTPGFGDQI NNDNCWDPIL GYINEQYEQY LQEEILITRQ RHIPDTRVHC CVYFVPPTGH CLRPLDIEFL QRLCRTVNVV PVIARADSLT MEEREAFRRR IQQNLRTHCI DVYPQMCFDE DINDKILNSK LRDRIPFAVV GADQEHLVNG RCVLGRKTKW GIIEVENMAH CEFPLLRDLL IRSHLQDLKD ITHNIHYENY RVIRLNESHL LPRGPGWVNL APASPGQLTT PRTFKVCRGA HDDSDDEF
Septins, such as SEPT12, are conserved GTP-binding proteins that function as dynamic, regulatable scaffolds for the recruitment of other proteins. They are involved in membrane dynamics, vesicle trafficking, apoptosis, and cytoskeleton remodeling, as well as infection, neurodegeneration, and neoplasia (Hall et al., 2005 [PubMed 15915442]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Septin-12; testicular tissue protein Li 168
Gene Aliases: SEPT12; SEPTIN12; SPGF10
UniProt ID: (Human) Q8IYM1
Entrez Gene ID: (Human) 124404
Molecular Function: cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support