Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Immunogen sequence: ESFFTWFTDH SDAGADELGE VIKDDIWPNP LQYYLVPDMD DEEGEGEEDD DDDEEEEGLE DIDEEGDEDE GEEDEDDDEG EEGEEDEGED D
SET is a multitasking protein, involved in apoptosis, transcription, nucleosome assembly and histone binding. Isoform 2 anti-apoptotic activity is mediated by inhibition of the GZMA-activated DNase, NME1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HLA-DR-associated protein II; I IGAAD; I-2PP2A; IGAAD; Inhibitor of granzyme A-activated DNase; inhibitor-2 of protein phosphatase-2A; Liver regeneration-related protein LRRGR00002; OTTHUMP00000022286; PHAPII; Phosphatase 2A inhibitor I2PP2A; protein phosphatase type 2A inhibitor; Protein SET; RP11-216B9.3; SET nuclear oncogene; SET translocation; SET translocation (myeloid leukemia-associated); TAF-1a; TAF-1b; TAF-I; Template-activating factor I; Template-Activating Factor-I, chromatin remodelling factor
Gene Aliases: 2610030F17Rik; 2PP2A; 5730420M11Rik; AA407739; Ab1-115; I-2PP2A; I2PP2A; IGAAD; IPP2A2; PHAPII; SET; StF-IT-1; TAF-I; TAF-IBETA
UniProt ID: (Human) Q01105, (Mouse) Q9CY82, (Rat) Q63945
Entrez Gene ID: (Human) 6418, (Mouse) 56086, (Rat) 307947
Molecular Function: chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support