Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLQGPGSLL LLFLASHCCL GSARGLFLFG QPDFSYKRSN CKPIPVNLQL CHGIEYQNMR LPNLLGHETM KEVLEQAGAW IPLVMKQCHP DTKKFLCSLF APVCLDDLDE TIQPCHSLCV QVKDRCAPVM SAFGFPWPDM LECDRFPQDN DLCIPLASSD HLLPATEEAP KVCEACKNKN DDDNDIMETL CKNDFALKIK VKEITYINRD TKIILETKSK TIYKLNGVSE RDLKKSVLWL KDSLQCTCEE MNDINAPYLV MGQKQGGELV ITSVKRWQKG QREFKRISRS IRKLQC (1-295 aa encoded by B C008666)
This gene encodes a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. Methylation of this gene is a potential marker for the presence of colorectal cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FRP 2; FRP-2; MGC128911; MGC153618; MGC53344; SARP 1; SARP-1; SDF 5; secreted apoptosis related protein 1; Secreted apoptosis-related protein 1; Secreted frizzled-related protein 2; sFRP 2; sFRP-2; Sfrp2 protein; testicular tissue protein Li 170
Gene Aliases: FKSG12; FRP-2; FRP2; SARP1; SDF-5; SFRP2; UNQ361/PRO697
UniProt ID: (Human) Q96HF1
Entrez Gene ID: (Human) 6423, (Rat) 310552
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support