Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MSMLPSFGFT QEQVACVCEV LQQGGNLERL GRFLWSLPAC DHLHKNESVL KAKAVVAFHR GNFRELYKIL ESHQFSPHNH PKLQQLWLKA HYVEAEKLCG RPLGAVGKYR VRRKFPLPRT IWDGEETSYC FKEKSRGVLR EWYAHNPYPS PREKRELAEA TGLTTTQVSN WFKNRRQRDR AAEAKERENT ENNNSSSNKQ NQLSPLEGGK PLMSSSEEEF SPPQSPDQNS VLLLQGNMGH ARSSNYSLPG LTASQPSHGL QTHQHQLQDS LLGPLTSSLV DLGS
The protein encoded by this gene is a homeobox protein that is similar to the Drosophila 'sine oculis' gene product. This gene is found in a cluster of related genes on chromosome 14 and is thought to be involved in limb development. Defects in this gene are a cause of autosomal dominant deafness type 23 (DFNA23) and branchiootic syndrome type 3 (BOS3).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein SIX1; Sine oculis homeobox homolog 1
Gene Aliases: BOS3; DFNA23; SIX1; TIP39
UniProt ID: (Human) Q15475
Entrez Gene ID: (Human) 6495
Molecular Function: homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support