Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: QAIGPSGMRS LAEPGCPTHG SAESPSTAAS PTTSVSSLTE RADTGTSILS VTSSDSECDV
This gene encodes a member of the sine oculis homeobox transcription factor family. The encoded protein plays a role in eye development. Mutations in this gene have been associated with holoprosencephaly type 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein SIX3; Sine oculis homeobox homolog 3; sine oculis homeobox-like protein 3
Gene Aliases: HPE2; SIX3
UniProt ID: (Human) O95343
Entrez Gene ID: (Human) 6496
Molecular Function: homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support