Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MACRPRSPPR HQSRCDGDAS PPSPARWSLG RKRRADGRRW RPEDAEEAEH RGAERRPESF TTPEGPKPRS RCSDWASAVE EDEMRTRVNK EMARYKRKLL INDFGRERKS SSGSSDSKES MSTVPADFET DESVLMRRQK QINYGKNTIA YDRYIKEVPR HLRQPGIHPK TPNKFKKYSR RSWDQQIKLW KVALHFWDPP AEEGCDLQEI HPVDLESAES SSEPQTSSQD DFDVYSGTPT KVRHMDSQVE DEFDLEACLT EPLRDFSAMS
This gene encodes a protein that binds to the stem-loop structure in replication-dependent histone mRNAs. Histone mRNAs do not contain introns or polyadenylation signals, and are processed by endonucleolytic cleavage. The stem-loop structure is essential for efficient processing but this structure also controls the transport, translation and stability of histone mRNAs. Expression of the protein is regulated during the cell cycle, increasing more than 10-fold during the latter part of G1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hairpin binding protein, histone; histone binding protein; Histone RNA hairpin-binding protein; histone stem-loop binding protein; Histone stem-loop-binding protein; stem-loop (histone) binding protein
Gene Aliases: HBP; SLBP
UniProt ID: (Human) Q14493
Entrez Gene ID: (Human) 7884
Molecular Function: RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support