Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: RYQPEQPNLV YQMASTSDEL DPADQNELAS TNDSQLGFLP EAEMFSLKTI LSPKNMEPSK IS
SLC7A1 is a high-affinity, low capacity permease involved in the transport of the cationic amino acids (arginine, lysine and ornithine) in non-hepatic tissues. It may also function as an ecotropic retroviral leukemia receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: amino acid transporter, cationic 1; CAT-1; CAT1; Ecotropic retroviral leukemia receptor homolog; ecotropic retroviral receptor; Ecotropic retrovirus receptor homolog; High affinity cationic amino acid transporter 1; solute carrier family 7 (cationic amino acid transporter, y+ system), member 1; Solute carrier family 7 member 1; System Y+ basic amino acid transporter
Gene Aliases: ATRC1; CAT-1; ERR; HCAT1; REC1L; SLC7A1
UniProt ID: (Human) P30825
Entrez Gene ID: (Human) 6541
Molecular Function: amino acid transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support