Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RDEYKPLDLS DSTLSYTETE ATNSLITAPG EFSDASMSPD ATKPSHWCSV AYWEHRTRVG RLYAVYDQAV SIFYDLPQGS GFCLGQLNLE QRSESVRRTR
SMAD6 (mothers against decapentaplegic homolog 6) is an antagonist of signaling by TGF-beta (transforming growth factor) type 1 receptor superfamily members; has been shown to inhibit selectively BMP (bone morphogenetic proteins) signaling by competing with the co-SMAD SMAD4 for receptor-activated SMAD1. SMAD6 is an inhibitory SMAD (I-SMAD) or antagonistic SMAD.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MAD homolog 6; Mothers against decapentaplegic homolog 6; mothers against decapentaplegic, drosophila, homolog of, 6; SMAD; SMAD 6; SMAD family member 6; SMAD SMAD; SMAD, mothers against DPP homolog 6
Gene Aliases: AOVD2; HsT17432; MADH6; MADH7; SMAD6
UniProt ID: (Human) O43541
Entrez Gene ID: (Human) 4091
Molecular Function: DNA-binding transcription factor gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support