Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: LGKFDEPLDM SLFSFIGSPR INATGQGVTI FYVDRLGYYP YIDSITGVTV NGGIPQKISL QDHLDKAKKD ITFYMPVDNL GMAVIDWEEW RPTWARNWKP KDVYKNRSIE LVQQQNVQLS LTEATEKAKQ EFEKAGKDFL VETIKLGKLL RPNHLWGYYL FPD
Hyaluronidase degrades hyaluronic acid, a major structural proteoglycan found in extracellular matrices and basement membranes. Six members of the hyaluronidase family are clustered into two tightly linked groups on chromosome 3p21.3 and 7q31.3. This gene was previously referred to as HYAL1 and HYA1 and has since been assigned the official symbol SPAM1; another family member on chromosome 3p21.3 has been assigned HYAL1. This gene encodes a GPI-anchored enzyme located on the human sperm surface and inner acrosomal membrane. This multifunctional protein is a hyaluronidase that enables sperm to penetrate through the hyaluronic acid-rich cumulus cell layer surrounding the oocyte, a receptor that plays a role in hyaluronic acid induced cell signaling, and a receptor that is involved in sperm-zona pellucida adhesion. Abnormal expression of this gene in tumors has implicated this protein in degradation of basement membranes leading to tumor invasion and metastasis. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory sperm binding protein Li 96n; Hyal-PH20; Hyaluronidase PH-20; Hyaluronoglucosaminidase PH-20; MGC26532; PH-20 hyaluronidase; Sperm adhesion molecule 1; sperm adhesion molecule 1 (PH-20 hyaluronidase, zona pellucida binding); Sperm surface antigen 2B1; Sperm surface protein PH-20
Gene Aliases: AV039194; HEL-S-96n; HYA1; HYAL1; HYAL3; HYAL5; PH-20; PH20; SPAG15; Spam; SPAM1
UniProt ID: (Human) P38567, (Rat) Q62803, (Mouse) P48794
Entrez Gene ID: (Human) 6677, (Rat) 117037, (Mouse) 20690
Molecular Function: glycosidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support