Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 5
Immunogen sequence: GGGGGGGAPR GRYGPPSRRS ENRVVVSGLP PSGSWQDLKD HMREAGDVCY ADVYRDGTGV VEFVRKEDMT YAVRKLDNTK FRSHEGETAY IRVKVDGPRS P
UBE2E2 catalyzes the covalent attachment of ubiquitin to other proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alternative-splicing factor 1; ASF-1; ASF/SF2; pre-mRNA-splicing factor SF2, P33 subunit; Pre-mRNA-splicing factor SRp30a; Serine/arginine-rich splicing factor 1; splicing factor 2; Splicing factor, arginine/serine-rich 1; splicing factor, arginine/serine-rich 1 (ASF/SF2); splicing factor, arginine/serine-rich 1B; splicing factor, arginine/serine-rich, 30-KD, A; SR splicing factor 1
Gene Aliases: 1110054N12Rik; 5730507C05Rik; 6330415C05Rik; AI482334; ASF; AW491331; OK/SW-cl.3; SF2; SF2P33; SFRS1; SRp30a; SRSF1
UniProt ID: (Human) Q07955, (Mouse) Q6PDM2
Entrez Gene ID: (Human) 6426, (Rat) 689890, (Mouse) 110809
Molecular Function: RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support