Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Immunocytochemistry (ICC/IF) |
1:10-1:50 | - |
ELISA (ELISA) |
1:20-1:200 | - |
Product Specifications | |
---|---|
Species Reactivity |
Bacteria |
Host/Isotype |
Mouse / IgG3 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
A68L |
Immunogen |
Strep pneumoniae native protein |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
100 µg/mL |
Storage conditions |
4° C |
RRID |
AB_2539635 |
This antibody is reactive with the S. pneumoniae common C-polysaccharide (backbone epitope).
Purity is >90%.
There are more than 85 antigenic types of S. pneumoniae, which are determined by capsule antigens. Altough S. pneumoniae is part of the normal respiratory flora, invasion commonly results in pneumonia as well as meningitis. S. pneumoniae is protected from phagocytosis by its polysaccharide capsule which is also a prominent virulence factor. Infections can usually be treated with antibiotics, although resistant strains are emerging.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Pneumococcus; s pneumoniae; S. pneumoniae
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support