Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCP11L2 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: t-complex 11, testis-specific-like 2; T-complex protein 11-like protein 2
Gene Aliases: TCP11L2
UniProt ID: (Human) Q8N4U5
Entrez Gene ID: (Human) 255394
Molecular Function: microtubule or microtubule-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support