Search Thermo Fisher Scientific
MA1-90998 detects TCR V beta 16.1 from rat samples.
MA1-90998 has been successfully used in immunohistochemistry (frozen tissue), immunoprecipitation applications.
The MA1-90998 immunogen is: PVG RT7.2 Rat Thymocytes