Product References
Substrate Stiffness-Driven Membrane Tension Modulates Vesicular Trafficking via Caveolin-1.
ACS nano
Lachowski D,Matellan C,Gopal S,Cortes E,Robinson BK,Saiani A,Miller AF,Stevens MM,Del Río Hernández AE
MA5-13688 was used in Immunohistochemistry-immunofluorescence to show that TIMP-1 and caveolin-1 expression increases in cirrhosis and hepatocellular carcinoma and position stiffness-dependent membrane tension as a key regulator of enzyme secretion and function and a potential target for therapeutic strategies that aim at modulating ECM remodeling in chronic liver disease.
Tue Mar 22 00:00:00 EDT 2022
Substrate Stiffness-Driven Membrane Tension Modulates Vesicular Trafficking via Caveolin-1.
ACS nano
Lachowski D,Matellan C,Gopal S,Cortes E,Robinson BK,Saiani A,Miller AF,Stevens MM,Del Río Hernández AE
MA5-13688 was used in Immunohistochemistry-immunofluorescence to show that TIMP-1 and caveolin-1 expression increases in cirrhosis and hepatocellular carcinoma and position stiffness-dependent membrane tension as a key regulator of enzyme secretion and function and a potential target for therapeutic strategies that aim at modulating ECM remodeling in chronic liver disease.
Tue Mar 22 00:00:00 EDT 2022
Substrate Stiffness-Driven Membrane Tension Modulates Vesicular Trafficking via Caveolin-1.
ACS nano
Lachowski D,Matellan C,Gopal S,Cortes E,Robinson BK,Saiani A,Miller AF,Stevens MM,Del Río Hernández AE
MA5-13688 was used in Immunohistochemistry-immunofluorescence to show that TIMP-1 and caveolin-1 expression increases in cirrhosis and hepatocellular carcinoma and position stiffness-dependent membrane tension as a key regulator of enzyme secretion and function and a potential target for therapeutic strategies that aim at modulating ECM remodeling in chronic liver disease.
Tue Mar 22 00:00:00 EDT 2022
Cancer Burden Is Controlled by Mural Cell-β3-Integrin Regulated Crosstalk with Tumor Cells.
Cell
Wong PP,Muñoz-Félix JM,Hijazi M,Kim H,Robinson SD,De Luxán-Delgado B,Rodríguez-Hernández I,Maiques O,Meng YM,Meng Q,Bodrug N,Dukinfield MS,Reynolds LE,Elia G,Clear A,Harwood C,Wang Y,Campbell JJ,Singh R,Zhang P,Schall TJ,Matchett KP,Henderson NC,Szlosarek PW,Dreger SA,Smith S,Jones JL,Gribben JG,Cutillas PR,Meier P,Sanz-Moreno V,Hodivala-Dilke KM
MA5-13688 was used in Western Blot to investigate if mural cells can control tumor growth via paracrine signals regulated by β3-integrin.
Thu Jun 11 00:00:00 EDT 2020
Pulsed focused ultrasound enhances the therapeutic effect of mesenchymal stromal cell-derived extracellular vesicles in acute kidney injury.
Stem cell research & therapy
Ullah M,Liu DD,Rai S,Razavi M,Concepcion W,Thakor AS
MA5-13688 was used in Western Blotting to show that pFUS may be a strategy for enhancing the therapeutic efficacy of MSC-derived EV treatment for the treatment of AKI.
Mon Sep 14 00:00:00 EDT 2020
Pulsed focused ultrasound enhances the therapeutic effect of mesenchymal stromal cell-derived extracellular vesicles in acute kidney injury.
Stem cell research & therapy
Ullah M,Liu DD,Rai S,Razavi M,Concepcion W,Thakor AS
MA5-13688 was used in Western Blotting to show that pFUS may be a strategy for enhancing the therapeutic efficacy of MSC-derived EV treatment for the treatment of AKI.
Mon Sep 14 00:00:00 EDT 2020
MAPKAP Kinase-2 Drives Expression of Angiogenic Factors by Tumor-Associated Macrophages in a Model of Inflammation-Induced Colon Cancer.
Frontiers in immunology
Suarez-Lopez L,Kong YW,Sriram G,Patterson JC,Rosenberg S,Morandell S,Haigis KM,Yaffe MB
MA5-13688 was used in Immunohistochemistry to identify a role for MK2-dependent regulation of the well-known pro-angiogenesis factor CXCL-12/SDF-1 secreted by tumor associated-macrophages, in addition to MK2-dependent regulation of Serpin-E1/PAI-1 by several cell types within the tumor microenvironment.
Wed Jun 23 00:00:00 EDT 2021
MAPKAP Kinase-2 Drives Expression of Angiogenic Factors by Tumor-Associated Macrophages in a Model of Inflammation-Induced Colon Cancer.
Frontiers in immunology
Suarez-Lopez L,Kong YW,Sriram G,Patterson JC,Rosenberg S,Morandell S,Haigis KM,Yaffe MB
MA5-13688 was used in Immunohistochemistry to identify a role for MK2-dependent regulation of the well-known pro-angiogenesis factor CXCL-12/SDF-1 secreted by tumor associated-macrophages, in addition to MK2-dependent regulation of Serpin-E1/PAI-1 by several cell types within the tumor microenvironment.
Wed Jun 23 00:00:00 EDT 2021
EphrinA1-Fc Attenuates Ventricular Remodeling and Dysfunction in Chronically Nonreperfused WT but not EphA2-R-M mice.
International journal of molecular sciences
Whitehurst KS,Chan VA,Estes HK,Valsaraj S,Kent S,Sharma UM,Chase RC,Bhuiyan M,Virag JAI
Published figure using TIMP1 monoclonal antibody (Product # MA5-13688) in Western Blot
Thu Aug 13 00:00:00 EDT 2020
The Activity of Matrix Metalloproteinases (MMP-2, MMP-9) and Their Tissue Inhibitors (TIMP-1, TIMP-3) in the Cerebral Cortex and Hippocampus in Experimental Acanthamoebiasis.
International journal of molecular sciences
Łanocha-Arendarczyk N,Baranowska-Bosiacka I,Gutowska I,Kolasa-Wołosiuk A,Kot K,Łanocha A,Metryka E,Wiszniewska B,Chlubek D,Kosik-Bogacka D
MA5-13688 was used in Immunohistochemistry to determine whether Acanthamoeba species may affect the levels of matrix metalloproteinases(MMPs)-2/-9, their tissue inhibitors, and MMP-9/TIMP-1, MMP-2/TIMP-3 ratios in the cerebral cortex and hippocampus, in relation to the host's immunological status.
Wed Dec 19 00:00:00 EST 2018
Expression and Clinical Significance of Metalloproteases and Their Inhibitors by Endothelial Cells From Invasive Breast Carcinomas.
Clinical breast cancer
Cid S,Eiro N,González LO,Beridze N,Vazquez J,Vizoso FJ
MA5-13688 was used in immunohistochemistry - paraffin section to study the clinical significance and expression of metalloproteases and their inhibitors in invasive breast carcinoma endothelial cells
Mon Aug 01 00:00:00 EDT 2016
The Predictive and Prognostic Role of Topoisomerase IIα and Tissue Inhibitor of Metalloproteinases 1 Expression in Locally Advanced Breast Carcinoma of Egyptian Patients Treated With Anthracycline-based Neoadjuvant Chemotherapy.
Applied immunohistochemistry & molecular morphology : AIMM
El Rebey HS,Aiad HA,Abulkheir IL,Asaad NY,El-Wahed MM,Abulkasem FM,Mahmoud SF
MA5-13688 was used in immunohistochemistry to determine the prognostic and predictive role of topoisomerase IIalpha and tissue inhibitor of metalloproteinases 1 in patient with locally advanced breast cancer after anthracycline-based neoadjuvant chemotherapy
Tue Mar 01 00:00:00 EST 2016
Molecular events underlying interleukin-6 independence in a subclone of the CMA-03 multiple myeloma cell line.
Genes, chromosomes & cancer
Verdelli D,Nobili L,Todoerti K,Mosca L,Fabris S,D'Anca M,Pellegrino E,Piva R,Inghirami G,Capelli C,Introna M,Baldini L,Chiaramonte R,Lombardi L,Neri A
MA5-13688 was used in western blot to study the molecular basis for the IL-6-independent growth of a variant of the CMA-03 multiple myeloma cell line
Sat Feb 01 00:00:00 EST 2014
Expression of matrix metalloproteinases and their inhibitors in different immunohistochemical-based molecular subtypes of breast cancer.
BMC cancer
Kim GE,Lee JS,Choi YD,Lee KH,Lee JH,Nam JH,Choi C,Kim SS,Park MH,Yoon JH,Kweon SS
MA5-13688 was used in Immunohistochemistry to assess the prevalence and clinical significance of matrix metalloproteinase and tissue inhibitor of metalloproteinase expression in invasive breast cancer to determine its association with immunohistochemical-based molecular classification.
Tue Dec 16 00:00:00 EST 2014
Correlation of bacterial coinfection versus matrix metalloproteinase 9 and tissue inhibitor of metalloproteinase 1 expression in aortic aneurysm and atherosclerosis.
Annals of vascular surgery
Roggério A,Sambiase NV,Palomino SA,de Castro MA,da Silva ES,Stolf NG,de Lourdes Higuchi M
MA5-13688 was used in immunohistochemistry - paraffin section to look for an association between Chlamydophila pneumoniae, Mycoplasma pneumoniae, matrix metalloproteinase 9, and tissue inhibitor of metalloproteinase 1 in aneurysmatic atherosclerotic lesions, and atherosclerotic nonaneurysmatic lesions
Tue Oct 01 00:00:00 EDT 2013
Comparative study of the immunohistochemical expression of tissue inhibitors of metalloproteinases 1 and 2 between clearly invasive carcinomas and "in situ" trophoblast invasion.
Medical oncology (Northwood, London, England)
Dimo B,Ioannidis I,Karameris A,Vilaras G,Tzoumakari P,Nonni A,Patsouris E,Lazaris AC
MA5-13688 was used in immunohistochemistry to compare the immunohistochemical expression of TIMP1 and TIMP2 between clearly invasive carcinomas and "in situ" trophoblast invasion
Sat Sep 01 00:00:00 EDT 2012
Immunoexpression of matrix metalloproteinases and their inhibitors in different areas of oral squamous cell carcinoma.
Australian dental journal
Suarez-Roa ML,Asbun-Bojalil J,Ruiz-Godoy LM,Meneses-García AA
MA5-13688 was used in immunohistochemistry to study the expression of MMPs and TIMPs in different histopathological regions of oral squamous cell carcinomas
Sat Sep 01 00:00:00 EDT 2012
Study of matrix metalloproteinases and their inhibitors in prostate cancer.
British journal of cancer
Escaff S,Fernández JM,González LO,Suárez A,González-Reyes S,González JM,Vizoso FJ
MA5-13688 was used in immunohistochemistry to study the expression of MMPs and TIMPs in prostate cancer
Tue Mar 02 00:00:00 EST 2010
Hypoxia promotes metastasis in human gastric cancer by up-regulating the 67-kDa laminin receptor.
Cancer science
Liu L,Sun L,Zhao P,Yao L,Jin H,Liang S,Wang Y,Zhang D,Pang Y,Shi Y,Chai N,Zhang H,Zhang H
MA5-13688 was used in western blot to study the role of the 67 kDa laminin receptor in the mechanism by which hypoxia promotes human gastric cancer metastasis
Thu Jul 01 00:00:00 EDT 2010
Expression of metalloproteases and their inhibitors by tumor and stromal cells in ductal carcinoma in situ of the breast and their relationship with microinvasive events.
Journal of cancer research and clinical oncology
González LO,González-Reyes S,Junquera S,Marín L,González L,Del Casar JM,González JM,Vizoso F
MA5-13688 was used in immunohistochemistry to study the immunohistochemical expression of MMPs and TIMPs in ductal carcinoma in situ of the breast
Wed Sep 01 00:00:00 EDT 2010
Expression of metalloproteases and their inhibitors in primary tumors and in local recurrences after mastectomy for breast cancer.
Journal of cancer research and clinical oncology
del Casar JM,Carreño G,González LO,Junquera S,González-Reyes S,González JM,Bongera M,Merino AM,Vizoso FJ
MA5-13688 was used in immunohistochemistry to study the expression of MMPs and TIMPs in breast cancer primary tumors and in local recurrences following mastectomy
Thu Jul 01 00:00:00 EDT 2010
Expression of metalloproteases and their inhibitors in different histological types of breast cancer.
Journal of cancer research and clinical oncology
Del Casar JM,González-Reyes S,González LO,González JM,Junquera S,Bongera M,García MF,Andicoechea A,Serra C,Vizoso FJ
MA5-13688 was used in immunohistochemistry to study the expression of MMPs and TIMPs in different histological subtypes of breast cancer
Tue Jun 01 00:00:00 EDT 2010
Immunohistochemical study of matrix metalloproteinases and their inhibitors in pure and mixed invasive and in situ ductal carcinomas of the breast.
Human pathology
Gonzalez LO,Junquera S,del Casar JM,González L,Marín L,González-Reyes S,Andicoechea A,González-Fernández R,González JM,Pérez-Fernández R,Vizoso FJ
MA5-13688 was used in immunohistochemistry to study the immunohistochemical expression of MMPs and TIMPs in differerent types of breast carcinoma
Thu Jul 01 00:00:00 EDT 2010
Immunohistochemical expression of matrix metalloproteinases in photodamaged skin by photodynamic therapy.
The British journal of dermatology
Almeida Issa MC,Piñeiro-Maceira J,Farias RE,Pureza M,Raggio Luiz R,Manela-Azulay M
MA5-13688 was used in immunohistochemistry to study effect of photodynamic therapy on the expression of MMPs by photo-damaged skin
Tue Sep 01 00:00:00 EDT 2009
Menstrual activity of matrix metalloproteinases is decreased in endometrium regenerating after thermal ablation.
Human reproduction (Oxford, England)
Brun JL,Galant C,Delvaux D,Lemoine P,Henriet P,Courtoy PJ,Marbaix E
MA5-13688 was used in immunohistochemistry to examine the alteration of matrix metalloproteinases during mentrual cycle
Sun Feb 01 00:00:00 EST 2009
Comparative analysis and clinical value of the expression of metalloproteases and their inhibitors by intratumor stromal fibroblasts and those at the invasive front of breast carcinomas.
Breast cancer research and treatment
Del Casar JM,González LO,Alvarez E,Junquera S,Marín L,González L,Bongera M,Vázquez J,Vizoso FJ
MA5-13688 was used in immunohistochemistry to study the clinical significance MMP and TIMP expression by intratumor stromal and invasive front fibroblasts in breast carcinoma
Wed Jul 01 00:00:00 EDT 2009
A mouse to human search for plasma proteome changes associated with pancreatic tumor development.
PLoS medicine
Faca VM,Song KS,Wang H,Zhang Q,Krasnoselsky AL,Newcomb LF,Plentz RR,Gurumurthy S,Redston MS,Pitteri SJ,Pereira-Faca SR,Ireton RC,Katayama H,Glukhova V,Phanstiel D,Brenner DE,Anderson MA,Misek D,Scholler N,Urban ND,Barnett MJ,Edelstein C,Goodman GE,Thornquist MD,McIntosh MW,DePinho RA,Bardeesy N,Hanash SM
MA5-13688 was used in immunohistochemistry to identify potential serum markers of pancreatic cancer using a proteomic analysis of a genetically engineered murine model
Tue Jun 10 00:00:00 EDT 2008
Serous and mucinous ovarian tumors express different profiles of MMP-2, -7, -9, MT1-MMP, and TIMP-1 and -2.
International journal of oncology
Brun JL,Cortez A,Commo F,Uzan S,Rouzier R,Daraï E
MA5-13688 was used in immunohistochemistry to investigate the expression of MMP-2,-7,-9,MT1-MMP, and TIMP-1 and -2 in ovarian tumors
Mon Dec 01 00:00:00 EST 2008
Gelatinase B expression as a prognostic factor in patients with stage II/III rectal carcinoma treated by postoperative adjuvant therapy.
American journal of clinical oncology
Unsal D,Akyurek N,Uner A,Erpolat OP,Han U,Akmansu M,Mentes BB,Dursun A
MA5-13688 was used in immunohistochemistry to investigate the prognostic value of gelatinase expression in patients with rectal carcinoma
Fri Feb 01 00:00:00 EST 2008
Overexpression of matrix metalloproteinases and their inhibitors in mononuclear inflammatory cells in breast cancer correlates with metastasis-relapse.
British journal of cancer
González LO,Pidal I,Junquera S,Corte MD,Vázquez J,Rodríguez JC,Lamelas ML,Merino AM,García-Muñiz JL,Vizoso FJ
Published figure using TIMP1 monoclonal antibody (Product # MA5-13688) in Immunohistochemistry
Mon Oct 08 00:00:00 EDT 2007
Overexpression of matrix metalloproteinases and their inhibitors in mononuclear inflammatory cells in breast cancer correlates with metastasis-relapse.
British journal of cancer
González LO,Pidal I,Junquera S,Corte MD,Vázquez J,Rodríguez JC,Lamelas ML,Merino AM,García-Muñiz JL,Vizoso FJ
Published figure using TIMP1 monoclonal antibody (Product # MA5-13688) in Immunohistochemistry
Mon Oct 08 00:00:00 EDT 2007
Study of matrix metalloproteinases and their inhibitors in breast cancer.
British journal of cancer
Vizoso FJ,González LO,Corte MD,Rodríguez JC,Vázquez J,Lamelas ML,Junquera S,Merino AM,García-Muñiz JL
MA5-13688 was used in immunohistochemistry to study the role of MMPs and TIMPs in breast cancer
Mon Mar 26 00:00:00 EDT 2007
Study of matrix metalloproteinases and their inhibitors in breast cancer.
British journal of cancer
Vizoso FJ,González LO,Corte MD,Rodríguez JC,Vázquez J,Lamelas ML,Junquera S,Merino AM,García-Muñiz JL
MA5-13688 was used in immunohistochemistry to study the role of MMPs and TIMPs in breast cancer
Mon Mar 26 00:00:00 EDT 2007
Matrix metalloproteinase-9 expression correlated with tumor response in patients with locally advanced rectal cancer undergoing preoperative chemoradiotherapy.
International journal of radiation oncology, biology, physics
Unsal Kilic D,Uner A,Akyurek N,Erpolat P,Dursun A,Pak Y
MA5-13688 was used in immunohistochemistry to examine the prognostic significance of matrix metalloproteinase-9 for locally advanced rectal cancer undergoing preoperative chemoradiotherapy
Mon Jan 01 00:00:00 EST 2007
Multinucleate giant cells release functionally unopposed matrix metalloproteinase-9 in vitro and in vivo.
The Journal of infectious diseases
Zhu XW,Price NM,Gilman RH,Recarvarren S,Friedland JS
MA5-13688 was used in immunohistochemistry to study the secretion of MMP-9 by multinucleate giant cells in vitro and in vivo
Mon Oct 01 00:00:00 EDT 2007
Matrix metalloproteinases and their inhibitors in the chamber angle of normal eyes and patients with primary open-angle glaucoma and exfoliation glaucoma.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie
Rönkkö S,Rekonen P,Kaarniranta K,Puustjärvi T,Teräsvirta M,Uusitalo H
MA5-13688 was used in immunohistochemistry to study the expression of MMPs and TIMPs in the chamber angle of normal eyes and patients with primary open-angle glaucoma and exfoliation glaucoma
Tue May 01 00:00:00 EDT 2007
Usefulness of MMP-9/TIMP-1 in predicting tumor recurrence in patients undergoing curative surgical resection for gastric carcinoma.
Digestive diseases and sciences
Seo YS,Park JJ,Kim JH,Kim JY,Yeon JE,Kim JS,Byun KS,Bak YT
MA5-13688 was used in immunohistochemistry to value of tumor MMP9 and TIMP1 levels for predicting tumor recurrence in patients undergoing curative resection for gastric carcinoma
Thu Mar 01 00:00:00 EST 2007
Tissue inhibitor of metalloproteinase-1 protects human breast epithelial cells from extrinsic cell death: a potential oncogenic activity of tissue inhibitor of metalloproteinase-1.
Cancer research
Liu XW,Taube ME,Jung KK,Dong Z,Lee YJ,Roshy S,Sloane BF,Fridman R,Kim HR
MA5-13688 was used in western blot to study the mechanisms by which TIMP-1 protects human breast epithelial cells from cell death
Tue Feb 01 00:00:00 EST 2005
In vitro modeling of human pancreatic duct epithelial cell transformation defines gene expression changes induced by K-ras oncogenic activation in pancreatic carcinogenesis.
Cancer research
Qian J,Niu J,Li M,Chiao PJ,Tsao MS
MA5-13688 was used in immunohistochemistry to investigate the role of K-ras oncogenic activation in pancreatic carcinogenesis
Wed Jun 15 00:00:00 EDT 2005
Upregulation of the tissue inhibitor of metalloproteinase-1 protein is associated with progression of human non-small-cell lung cancer.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Aljada IS,Ramnath N,Donohue K,Harvey S,Brooks JJ,Wiseman SM,Khoury T,Loewen G,Slocum HK,Anderson TM,Bepler G,Tan D
MA5-13688 was used in immunohistochemistry to investigate the association of TIMP-1 upregulation with the progression of human non-small-cell lung cancer
Sun Aug 15 00:00:00 EDT 2004
Tissue inhibitor of matrix metalloproteinase-1 overexpression in M1 myeloblasts impairs IL-6-induced differentiation.
Oncogene
Haviernik P,Lahoda C,Bradley HL,Hawley TS,Ramezani A,Hawley RG,Stetler-Stevenson M,Stetler-Stevenson WG,Bunting KD
MA5-13688 was used in blocking or activating experiment to study the role of TIMP1 overexpression in impairing IL-6-induced differentiation in M1 myeloblasts
Thu Dec 09 00:00:00 EST 2004
Systemic arterial expression of matrix metalloproteinases 2 and 9 in acute Kawasaki disease.
Arteriosclerosis, thrombosis, and vascular biology
Gavin PJ,Crawford SE,Shulman ST,Garcia FL,Rowley AH
MA5-13688 was used in immunohistochemistry to study the expression of MMP2, MMP9, TIMP1 and TIMP2 in arteries from patients with fatal acute Kawasaki's disease
Tue Apr 01 00:00:00 EST 2003
Abnormal expression of matrix metalloproteinases and tissue inhibitors of metalloproteinases in brain arteriovenous malformations.
Stroke
Hashimoto T,Wen G,Lawton MT,Boudreau NJ,Bollen AW,Yang GY,Barbaro NM,Higashida RT,Dowd CF,Halbach VV,Young WL
MA5-13688 was used in western blot to study the expression of MMPs and TIMPs in brain arteriovenous malformations
Tue Apr 01 00:00:00 EST 2003
The in vitro effects of dehydroepiandrosterone on human osteoarthritic chondrocytes.
Osteoarthritis and cartilage
Jo H,Park JS,Kim EM,Jung MY,Lee SH,Seong SC,Park SC,Kim HJ,Lee MC
MA5-13688 was used in western blot to study the effects of dehydroepiandrosterone on chondrocytes isolated from human osteoarthritic cartilage
Fri Aug 01 00:00:00 EDT 2003
Unopposed matrix metalloproteinase-9 expression in human tuberculous granuloma and the role of TNF-alpha-dependent monocyte networks.
Journal of immunology (Baltimore, Md. : 1950)
Price NM,Gilman RH,Uddin J,Recavarren S,Friedland JS
MA5-13688 was used in immunohistochemistry to investigate the roles of metalloproteinase-9 and TNF-alpha-dependent monocyte networks in human tuberculous granulom
Sat Nov 15 00:00:00 EST 2003
Imbalance between expression of matrix metalloproteinase-9 and tissue inhibitor of metalloproteinase-1 in invasiveness and metastasis of human gastric carcinoma.
World journal of gastroenterology
Zhang S,Li L,Lin JY,Lin H
MA5-13688 was used in immunohistochemistry to study the role of MMP-9 and TIMP-1 imbalance in metastasis and poor prognosis in gastric carcinoma
Thu May 01 00:00:00 EDT 2003
Tissue inhibitor of metalloproteinase-1 protects human breast epithelial cells against intrinsic apoptotic cell death via the focal adhesion kinase/phosphatidylinositol 3-kinase and MAPK signaling pathway.
The Journal of biological chemistry
Liu XW,Bernardo MM,Fridman R,Kim HR
MA5-13688 was used in western blot to study the mechanism by which TIMP1 protects human breast epithelial cells against intrinsic apoptotic cell death
Fri Oct 10 00:00:00 EDT 2003