Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
CAND2 may play a role in the assembly of ubiquitin ligase complexes and modulate the ubiquitination of target proteins. And it may be also a transcription regulator.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cullin-associated and neddylation-dissociated 2 (putative); Cullin-associated and neddylation-dissociated protein 2; Cullin-associated NEDD8-dissociated protein 2; Epididymis tissue protein Li 169; p120 CAND2; TBP interacting protein; TBP-interacting protein 120B; TBP-interacting protein b; TBP-interacting protein of 120 kDa B
Gene Aliases: 2210404G23Rik; CAND2; KIAA0667; mKIAA0667; TIP120B; Tp120b
UniProt ID: (Human) O75155, (Mouse) Q6ZQ73
Entrez Gene ID: (Human) 23066, (Mouse) 67088
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support