Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: KKETCKLVCT KTYHTEKAED KQKLEFLKKS MLLNYQHHWI VDNMPVTWCY DVEDGQRFCN PGFPIGCYIT DKGHAKDACV ISSDFHERDT FYIFNHVDIK
In the intracellular compartments, may function as a channel or small molecule transporter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 76 kDa membrane protein; dinucleotide oxidase disulfide thiol exchanger 3 superfamily member 2; p76; Transmembrane 9 superfamily member 2; transmembrane protein 9 superfamily member 2
Gene Aliases: P76; TM9SF2
UniProt ID: (Human) Q99805
Entrez Gene ID: (Human) 9375
Molecular Function: transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support