Search Thermo Fisher Scientific
The immunogenic sequence shares 94% identity with TRPC3 and TRPC7 sequences.
Thought to form a receptor-activated non-selective calcium permeant cation channel. Probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. Activated by diacylglycerol (DAG) in a membrane-delimited fashion, independently of protein kinase C, and by inositol-1,4,5-triphosphate receptors (ITPR) with bound IP3. May also be activated by internal calcium store depletion. Abundantly expressed in brain. Concentrated in cerebellar Purkinje cells and sparsely localized in cerebellar granule lyer, pontine nuclei and thalamus. Lower levels detected in other tissues.