Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: RHEIKNSFKN NYEKALKQYN STGDYRSHAV DKIQNTLHCC GVTDYRDWTD TNYYSEKGFP KSCCKLEDCT PQRDADKVNN EGCFIKVMTI IESE
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein and is highly similar in sequence to the transmembrane 4 superfamily member 2. The use of alternate polyadenylation sites has been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A15 homolog; Putative NF-kappa-B-activating protein 321; T245 protein; tetraspan TM4SF; Tetraspanin TM4-D; Tetraspanin-6; Transmembrane 4 superfamily member 6; Tspan-6
Gene Aliases: T245; TM4SF6; TSPAN-6; TSPAN6; UNQ767/PRO1560
UniProt ID: (Human) O43657
Entrez Gene ID: (Human) 7105
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support