Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500 | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:150 | - |
Immunoprecipitation (IP) |
2 µg/500 µL | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
OTI4C8 |
Immunogen |
Full length human recombinant protein of human TYRO3 produced in HEK293T cell. |
Conjugate |
Unconjugated
|
Form |
lyophilized |
Concentration |
1 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS, pH 7.3, with 8% trehalose |
Contains |
no preservative |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
TYRO3 (tyrosine-protein kinase receptor TYRO3) is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding to several ligands including TULP1 or GAS6. It regulates many physiological processes including cell survival, migration, and differentiation. Ligand binding at the cell surface induces dimerization and autophosphorylation of TYRO3 on its intracellular domain that provides docking sites for downstream signaling molecules. Following activation by a ligand, TYRO3 plays a role in various processes such as neuron protection from excitotoxic injury, platelet aggregation, and cytoskeleton reorganization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ16467; soluble tyro 3; soluble tyro3; styro 3; styro3; tyro 3; Tyrosine-protein kinase BYK; Tyrosine-protein kinase DTK; Tyrosine-protein kinase receptor TYRO3; Tyrosine-protein kinase RSE; Tyrosine-protein kinase SKY; Tyrosine-protein kinase TIF
Gene Aliases: BYK; DTK; Etk-2; Rek; RSE; SKY; TIF; TYRO3
UniProt ID: (Human) Q06418
Entrez Gene ID: (Human) 7301
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support