Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PSSQLYEHTV EGGEKQVVFT HRINLPPSTG CGCPPGTEPP VLASEVQALR VRLEILEELV KGLKEQCTGG CCPASAQAGT GQTDVRTLCS LHGVFDLSRC TCSCEPGWGG PTCSDPTDA
Antigen sequence identity: Mouse 92%; Rat 47%
Tenascin-X (TNXB) is a extracellular matrix glycoprotein predominantly located in the outer reticular lamina of the basement membrane (BM). It interacts with many other ECM proteins and it accelerates collagen fibrillogenesisin vitro. TNXB plays a role in interactions between cell and ECM, antiadhesive effect, inhibiting cell migration, and in maintaining homeostasis of the extracellular matrix. Deficiency of TNXB has been associated with the connective tissue disorder Ehlers-Danlos syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: growth-inhibiting protein 45; Hexabrachion-like protein; tenascin XB1; tenascin XB2; Tenascin-X; TN-X; TNXB2tenascin XB1; TNXBSTNXTENX; XB-S; XBTN-X
Gene Aliases: EDS3; HXBL; TENX; TN-X; TNX; TNXB; TNXB1; TNXB2; TNXBS; VUR8; XB; XBS
UniProt ID: (Human) P22105
Entrez Gene ID: (Human) 7148
Molecular Function:
extracellular matrix protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support