Search Thermo Fisher Scientific
MA3-217 detects both glycosylated and nonglycosylated thyroid stimulating hormone receptor (TSHR) from porcine tissues as well as recombinant human TSHR.
MA3-217 has been successfully used in Western blot, immunocytochemistry, and ELISA procedures. By Western blot, this antibody detects a 50 kDa and a 63 kDa protein representing unglycosylated and glycosylated TSRH respectively, in transfected Sf9 cell extract. By ELISA MA3-217 has been used in TSH competitive inhibition studies and to detect the presence of human TSHR.
The MA3-217 antigen is purified extracellular domain of recombinant human thyrotropin receptor. This antibody recognizes an epitope between amino acid residues 32-41 of human TSHR.
TSH Receptor gene is a membrane protein and a major controller of thyroid cell metabolism. The encoded protein is a receptor for thyrothropin and thyrostimulin, and its activity is mediated by adenylate cyclase. Defects in this gene are a cause of several types of hyperthyroidism. Three transcript variants encoding different isoforms have been found for this gene.