Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: class V beta-tubulin; tubulin beta; Tubulin beta class V; tubulin beta MGC4083; Tubulin beta-6 chain; tubulin, beta 6 class V
Gene Aliases: 2310057H16Rik; BB220206; HsT1601; TUBB-5; TUBB6
UniProt ID: (Human) Q9BUF5, (Mouse) Q922F4
Entrez Gene ID: (Dog) 480213, (Human) 84617, (Mouse) 67951
Molecular Function:
tubulin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support